Merck
  • Effects of ganoderic acid A on lipopolysaccharide-induced proinflammatory cytokine release from primary mouse microglia cultures.
Effects of ganoderic acid A on lipopolysaccharide-induced proinflammatory cytokine release from primary mouse microglia cultures.
Experimental and therapeutic medicine (2018-02-06)
Baojin Chi, Shuqiu Wang, Sheng Bi, Wenbo Qin, Dongmei Wu, Zhenguo Luo, Shiliang Gui, Dongwei Wang, Xingzhong Yin, Fangfang Wang
ABSTRACT
For several thousand years,
Sigma-Aldrich
Anti-IL6, affinity isolated antibody
Sigma-Aldrich
Anti-TNFA antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Interleukin-1β antibody produced in goat, IgG fraction of antiserum