Merck
  • Normal human adipose tissue functions and differentiation in patients with biallelic
Normal human adipose tissue functions and differentiation in patients with biallelic
Journal of lipid research (2017-10-08)
Michele Pelosi, Eric Testet, Soazig Le Lay, Isabelle Dugail, Xiaoyun Tang, Guillaume Mabilleau, Yamina Hamel, Marine Madrange, Thomas Blanc, Thierry Odent, Todd P W McMullen, Marco Alfò, David N Brindley, Pascale de Lonlay
ABSTRACT
Lipin-1 is a Mg