- Solid formulation of a supersaturable self-microemulsifying drug delivery system for valsartan with improved dissolution and bioavailability.
Solid formulation of a supersaturable self-microemulsifying drug delivery system for valsartan with improved dissolution and bioavailability.
Oncotarget (2017-12-08)
Dong Woo Yeom, Bo Ram Chae, Jin Han Kim, Jun Soo Chae, Dong Jun Shin, Chang Hyun Kim, Sung Rae Kim, Ji Ho Choi, Seh Hyon Song, Dongho Oh, Se Il Sohn, Young Wook Choi
PMID29212229
ABSTRACT
In order to improve the dissolution and oral bioavailability of valsartan (VST), and reduce the required volume for treatment, we previously formulated a supersaturable self-microemulsifying drug delivery system (SuSMEDDS) composed of VST (80 mg), Capmul