Merck
  • Effect of Genotype on the Sprouting of Pomegranate (Punica granatum L.) Seeds as a Source of Phenolic Compounds from Juice Industry by-Products.
Effect of Genotype on the Sprouting of Pomegranate (Punica granatum L.) Seeds as a Source of Phenolic Compounds from Juice Industry by-Products.
Plant foods for human nutrition (Dordrecht, Netherlands) (2017-11-04)
Beatrice Falcinelli, Ombretta Marconi, Stefano Maranghi, Stanley Lutts, Adolfo Rosati, Franco Famiani, Paolo Benincasa
ABSTRACT
Pomegranate (Punica granatum L.) fruits are used mainly by the juice industry, for which seeds are a by-product to be disposed of, though they could potentially be a source of bioactive compounds. In this work, germination (total germination percentage, G; mean germination time, MGT; time to reach 80% of germination, TG80; seedling shoot length, fresh weight and dry matter), and nutritional value (total phenolics, TP; total flavonoids, TF; total non-tannins, TNT; antioxidant activities) of pomegranate seeds and sprouts were determined on four commercial pomegranate cultivars (Akko, Dente di Cavallo, Mollar de Elche and Wonderful). Seeds were removed from ripe fruits and incubated in plastic trays containing sterile cotton wetted with distilled water. Sprout shoots were harvested when they reached the complete cotyledon expansion, i.e., the ready-to-eat stage. Akko showed the best germination performance (G = 98%; MGT = 14 days after sowing, DAS; TG80 = 16 DAS), followed by Mollar de Elche. Sprouting dramatically increased TP, TF, TNT and antioxidant activity in all genotypes, with the highest values recorded in Mollar de Elche and Dente di Cavallo. Overall, based on germination performance, Akko and Mollar de Elche would be the best cultivars for sprouting. Sprouting pomegranate seeds appears to be a suitable way of utilizing by-products of the juice industry to obtain bioactive compounds.