Merck
  • Preparative separation and quantitative determination of two kaurenoic acid isomers in root barks of Acanthopanax gracilistylus.
Preparative separation and quantitative determination of two kaurenoic acid isomers in root barks of Acanthopanax gracilistylus.
Chinese journal of natural medicines (2017-09-25)
Xin-Xin Xie, Ze-Jing Jiang, Zhi-Hong Cheng, Dao-Feng Chen
ABSTRACT
The kaurenoic acid-type diterpenoids in Acanthopanacis Cortex have been reported to be the major active components. However, the diterpenoids are present as position isomers that exacerbate the challenges in obtaining standards compounds. Little work has been done on the quantitative analysis of the diterpenoids in the herb. In the present study, two diterpenoid isomers ent-16βH,17-isovalerate-kauran-19-oic acid (1) and ent-16βH,17-methyl butanoate-kauran-19-oic acid (2) with high purity were separated by analytical HPLC, followed by recrystallization in acetone. Furthermore, an HPLC-ELSD method was developed and validated for simultaneous determination of 1 and 2 in 9 batches of Acanthopanacis Cortex samples. The HPLC separation and quantification was achieved in 40 min using an Agela Promosil C