Merck
  • CRISPR/Cas9 mediated G4946E substitution in the ryanodine receptor of Spodoptera exigua confers high levels of resistance to diamide insecticides.
CRISPR/Cas9 mediated G4946E substitution in the ryanodine receptor of Spodoptera exigua confers high levels of resistance to diamide insecticides.
Insect biochemistry and molecular biology (2017-09-16)
Yayun Zuo, Hui Wang, Yanjun Xu, Jianlei Huang, Shuwen Wu, Yidong Wu, Yihua Yang
ABSTRACT
Diamide insecticides selectively activate insect ryanodine receptors (RyRs), inducing uncontrolled release of calcium ions, and causing muscle contraction, paralysis and eventually death. The RyR
Product Description
Supelco
Cyantraniliprole, PESTANAL®, analytical standard
Pricing and availability is not currently available.