Merck
  • Rapid and orthogonal logic gating with a gibberellin-induced dimerization system.
Rapid and orthogonal logic gating with a gibberellin-induced dimerization system.
Nature chemical biology (2012-03-27)
Takafumi Miyamoto, Robert DeRose, Allison Suarez, Tasuku Ueno, Melinda Chen, Tai-ping Sun, Michael J Wolfgang, Chandrani Mukherjee, David J Meyers, Takanari Inoue
ABSTRACT
Using a newly synthesized gibberellin analog containing an acetoxymethyl group (GA(3)-AM) and its binding proteins, we developed an efficient chemically inducible dimerization (CID) system that is completely orthogonal to existing rapamycin-mediated protein dimerization. Combining the two systems should allow applications that have been difficult or impossible with only one CID system. By using both chemical inputs (rapamycin and GA(3)-AM), we designed and synthesized Boolean logic gates in living mammalian cells. These gates produced output signals such as fluorescence and membrane ruffling on a timescale of seconds, substantially faster than earlier intracellular logic gates. The use of two orthogonal dimerization systems in the same cell also allows for finer modulation of protein perturbations than is possible with a single dimerizer.
Sigma-Aldrich
GA3-AM, ≥95% (HPLC)
Pricing and availability is not currently available.