- Establishing an Artificial Pathway for De Novo Biosynthesis of Vanillyl Alcohol in Escherichia coli.
Establishing an Artificial Pathway for De Novo Biosynthesis of Vanillyl Alcohol in Escherichia coli.
ACS synthetic biology (2017-06-07)
Zhenya Chen, Xiaolin Shen, Jian Wang, Jia Wang, Ruihua Zhang, Justin Forrest Rey, Qipeng Yuan, Yajun Yan
PMID28586214
ABSTRACT
Vanillyl alcohol is a phenolic alcohol and is used as a flavoring agent in foods and beverages. In this paper, we propose a novel artificial pathway for microbial production of vanillyl alcohol from simple carbon sources. The pathway extends from 4-hydroxybenzoic acid (4-HBA), and needs only three heterologous enzymes, p-hydroxybenzoate hydroxylase (PobA), carboxylic acid reductase (CAR) and caffeate O-methyltransferase (COMT). First, we examined the promiscuous activity of COMT toward 3,4-dihydroxybenzyl alcohol and found a k