Merck
  • PRMT5 (Janus kinase-binding protein 1) catalyzes the formation of symmetric dimethylarginine residues in proteins.
PRMT5 (Janus kinase-binding protein 1) catalyzes the formation of symmetric dimethylarginine residues in proteins.
The Journal of biological chemistry (2001-06-20)
T L Branscombe, A Frankel, J H Lee, J R Cook, Z Yang, S Pestka, S Clarke
ABSTRACT
We have identified a new mammalian protein arginine N-methyltransferase, PRMT5, formerly designated Janus kinase-binding protein 1, that can catalyze the formation of omega-N(G)-monomethylarginine and symmetric omega-N(G),N(G')-dimethylarginine in a variety of proteins. A hemagglutinin peptide-tagged PRMT5 complex purified from human HeLa cells catalyzes the S-adenosyl-l-[methyl-(3)H]methionine-dependent in vitro methylation of myelin basic protein. When the radiolabeled myelin basic protein was acid-hydrolyzed to free amino acids, and the products were separated by high-resolution cation exchange chromatography, we were able to detect two tritiated species. One species co-migrated with a omega-N(G)-monomethylarginine standard, and the other co-chromatographed with a symmetric omega-N(G),N(G')-dimethylarginine standard. Upon base treatment, this second species formed methylamine, a breakdown product characteristic of symmetric omega-N(G),N(G')-dimethylarginine. Further analysis of these two species by thin layer chromatography confirmed their identification as omega-N(G)-monomethylarginine and symmetric omega-N(G),N(G')-dimethylarginine. The hemagglutinin-PRMT5 complex was also able to monomethylate and symmetrically dimethylate bovine histone H2A and a glutathione S-transferase-fibrillarin (amino acids 1-148) fusion protein (glutathione S-transferase-GAR). A mutation introduced into the S-adenosyl-l-methionine-binding motif I of a myc-tagged PRMT5 construct in COS-1 cells led to a near complete loss of observed enzymatic activity. PRMT5 is the first example of a catalytic chain for a type II protein arginine N-methyltransferase that can result in the formation of symmetric dimethylarginine residues as observed previously in myelin basic protein, Sm small nuclear ribonucleoproteins, and other polypeptides.
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)