Merck
  • Glomerulosclerosis Induced by Deficiency of Membrane-Associated Guanylate Kinase Inverted 2 in Kidney Podocytes.
Glomerulosclerosis Induced by Deficiency of Membrane-Associated Guanylate Kinase Inverted 2 in Kidney Podocytes.
Journal of the American Society of Nephrology : JASN (2017-05-26)
Naritoshi Shirata, Kan-Ichiro Ihara, Kanae Yamamoto-Nonaka, Takuto Seki, Shin-Ichi Makino, Juan Alejandro Oliva Trejo, Takafumi Miyake, Hiroyuki Yamada, Kirk Nicholas Campbell, Takahiko Nakagawa, Kiyoshi Mori, Motoko Yanagita, Peter Mundel, Katsuhiko Nishimori, Katsuhiko Asanuma
ABSTRACT
Membrane-associated guanylate kinase inverted 2 (MAGI-2) is a component of the slit diaphragm (SD) of glomerular podocytes. Here, we investigated the podocyte-specific function of MAGI-2 using newly generated podocyte-specific MAGI-2-knockout (MAGI-2-KO) mice. Compared with podocytes from wild-type mice, podocytes from MAGI-2-KO mice exhibited SD disruption, morphologic abnormalities of foot processes, and podocyte apoptosis leading to podocyte loss. These pathologic changes manifested as massive albuminuria by 8 weeks of age and glomerulosclerosis and significantly higher plasma creatinine levels at 12 weeks of age; all MAGI-2-KO mice died by 20 weeks of age. Loss of MAGI-2 in podocytes associated with decreased expression and nuclear translocation of dendrin, which is also a component of the SD complex. Dendrin translocates from the SD to the nucleus of injured podocytes, promoting apoptosis. Our coimmunoprecipitation and
Sigma-Aldrich
PP2, ≥98% (HPLC)