- Drop coating deposition Raman spectroscopy of proteinogenic amino acids compared with their solution and crystalline state.
Drop coating deposition Raman spectroscopy of proteinogenic amino acids compared with their solution and crystalline state.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-06-04)
Tomáš Pazderka, Vladimír Kopecký
PMID28577510
ABSTRACT
The Raman spectra of 20 proteinogenic amino acids were recorded in the solution, glass phase (as drop coating deposition Raman (DCDR) samples) and crystalline forms in the wide spectral range of 200-3200cm