Merck
  • Impaired endothelium-derived hyperpolarization-type relaxation in superior mesenteric arteries isolated from female Otsuka Long-Evans Tokushima Fatty rats.
Impaired endothelium-derived hyperpolarization-type relaxation in superior mesenteric arteries isolated from female Otsuka Long-Evans Tokushima Fatty rats.
European journal of pharmacology (2017-04-24)
Takayuki Matsumoto, Shota Kobayashi, Makoto Ando, Shun Watanabe, Maika Iguchi, Kumiko Taguchi, Tsuneo Kobayashi
ABSTRACT
Endothelium-derived hyperpolarization (EDH) is an important signaling mechanism of endothelium-dependent vasorelaxation, and little attention has been paid to the EDH-type responses in female metabolic syndrome such as that observed with type-2 diabetes. We previously reported that EDH-type relaxation was impaired in superior mesenteric arteries from male Otsuka Long-Evans Tokushima Fatty (OLETF) rat, a model of type-2 diabetes, however, the response was unclear in female OLETF rat. Thus, the aim of this study was to examine if EDH-type relaxation was altered in superior mesenteric arteries isolated from female OLETF rats compared to age-matched, control female Long-Evans Tokushima Otsuka (LETO) rats at age 50-59 weeks. We investigated concentration-relaxation curves for acetylcholine (at age 50-53 weeks), NS309 (an activator of small- and intermediate-conductance calcium-activated potassium channels) (at age 50-53 weeks), and GSK1016790A (an agonist of transient receptor potential vanilloid type 4, TRPV4) (at age 58 or 59 weeks) in the presence of the nitric oxide synthase inhibitor N
Sigma-Aldrich
NS309, ≥98% (HPLC), solid