Merck
  • Feasibility of a novel multispot nanoarray for antibiotic screening in honey.
Feasibility of a novel multispot nanoarray for antibiotic screening in honey.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-01-13)
Sara E McNamee, Giulia Rosar, Lidija Persic, Christopher T Elliott, Katrina Campbell
ABSTRACT
Practical solutions for multiple antibiotic determination in food are required by the food industry and regulators for cost-effective screening purposes. This study describes the feasibility in development and preliminary performance of a novel multispot nanoarray for antibiotic screening in honey. Using a multiplex approach, the metabolites of the four main nitrofuran antibiotics, including morpholinomethyl-2-oxazolidone (AMOZ), 3-amino-2-oxazolidinone (AOZ), semicarbazide (SEM), 1-aminohydantoin (AHD) and chloramphenicol (CAP), were simultaneously detected. Antibodies specific to the five antibiotics were nano-spotted onto microtitre plate wells and a direct competitive assay format was employed. The assay characteristics and performance were evaluated for feasibility as a screening tool for antibiotic determination in honey to replace traditional ELISAs. Optimisation of the spotting and assay parameters was undertaken with both individual and multiplex calibration curves generated in PBS and a honey matrix. The limits of detection as determined by the 20% inhibitory concentrations (IC
Product Description
Supelco
2-NP-AMOZ, VETRANAL®, analytical standard
Supelco
2-NP-AHD, VETRANAL®, analytical standard