Merck
Mitomycin C: small, fast and deadly (but very selective).
Chemistry & biology (1995-09-01)
M Tomasz
ABSTRACT
Mitomycin C, an important antitumor drug and antibiotic, has an extraordinary ability to crosslink DNA with high efficiency and absolute specificity for the sequence CpG. Recent results have shown how mitomycin C crosslinks DNA, and why the sequence specificity is so complete. This new understanding may allow the design of agents that mimic mitomycin C's economy of structure and can crosslink other sequences.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Roche
Mitomycin C, crystalline powder, 98%, pkg of 2 mg