Merck
  • Design and Synthesis of 4-Anilinothieno[2,3-d]pyrimidine-Based Compounds as Dual EGFR/HER-2 Inhibitors.
Design and Synthesis of 4-Anilinothieno[2,3-d]pyrimidine-Based Compounds as Dual EGFR/HER-2 Inhibitors.
Archiv der Pharmazie (2016-11-03)
Soha R Abd El Hadi, Deena S Lasheen, Mahmoud A Hassan, Khaled A M Abouzid
ABSTRACT
Dual inhibition of the epidermal growth factor receptor (EGFR) and the human epidermal growth factor receptor 2 (HER-2) is an attractive cancer therapeutic approach. In this study, new series of 4-anilinothieno[2,3-d]pyrimidines were designed, synthesized, and tested as dual EGFR/HER-2 kinase inhibitors. Five compounds (8a, 8b, 8e-g) demonstrated low to submicromolar inhibition of both kinases with IC