Merck
  • Cholesterol, phospholipid and phospholipase activity of ampullary and isthmic fluid from the bovine oviduct.
Cholesterol, phospholipid and phospholipase activity of ampullary and isthmic fluid from the bovine oviduct.
Journal of reproduction and fertility (1994-09-01)
A A Grippo, S H Anderson, D A Chapman, M A Henault, G J Killian
ABSTRACT
Cholesterol and phospholipid concentrations and phospholipase activity were measured in fluid from cannulae collected from the bovine oviductal isthmus and ampulla at different stages of the oestrous cycle. The cholesterol concentration and cholesterol normalized by protein were significantly (P = 0.03) greater in isthmic oviductal fluid (224.3 +/- 42.7 micrograms ml-1 over all stages) than in ampullary oviductal fluid (164.5 +/- 11.3 micrograms ml-1), and maximal concentrations (284.5 +/- 25.5 micrograms ml-1) were found during the luteal stage (serum progesterone concentration > or = 1.5 ng ml-1). The concentrations of the phospholipids sphingomyelin and lysophosphatidylcholine increased at different stages of the cycle and in different regions. In the ampulla, the concentration of sphingomyelin was significantly (P < 0.05) greater in oviductal fluid collected during the luteal phase (12.1 +/- 2.7% of total phospholipids) than in fluid collected near oestrus and ovulation (7.5 +/- 1.5% and 6.9 +/- 1%, respectively). The concentration of lysophosphatidylcholine was greater (P < 0.01) in ampullary (19.2 +/- 1.6% of total phospholipids) than in isthmic oviductal fluid (9.9 +/- 1.1%) collected near ovulation. The ratio of cholesterol to total phospholipid was highest in oviductal fluid collected from the isthmus during all stages (2.3 micrograms ml-1:% total phospholipid), while the minimal ratio was found in ampullary fluid collected near ovulation (1.5). Phospholipase activity was higher (P = 0.03) in isthmic oviductal fluid (20.4 +/- 3.2% product formed) than in ampullary oviductal fluid (14.6 +/- 1.4%); the lowest activity (12.6 +/- 1.7% product formed) was in fluid collected during the phase of the oestrous cycle immediately before ovulation.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein