Merck
  • Comparative microbiological studies of transcription inhibitors fidaxomicin and the rifamycins in Clostridium difficile.
Comparative microbiological studies of transcription inhibitors fidaxomicin and the rifamycins in Clostridium difficile.
Antimicrobial agents and chemotherapy (2014-02-20)
Farah Babakhani, Jaime Seddon, Pamela Sears
ABSTRACT
Fidaxomicin (FDX) is a narrow-spectrum antibiotic for the treatment of Clostridium difficile-associated diarrhea. While FDX and rifamycins share the same target (RNA polymerase), FDX exhibits a unique mode of action distinct from that of rifamycins. In comparative microbiological studies with C. difficile, FDX interacted synergistically with rifamycins, demonstrated a lower propensity for the development of resistance to rifamycins, and exhibited no cross-resistance with rifamycins. These results highlight differences in the mechanisms of action of FDX and rifamycins.
Sigma-Aldrich
Fidaxomicin, ≥95% (HPLC)