MilliporeSigma
Bartoli indole synthesis on solid supports.
Organic letters (2003-08-02)
Kerstin Knepper, Stefan Bräse
ABSTRACT
[reaction: see text] Bartoli indole synthesis has been performed for the first time on solid supports. Starting from Merrifield resin, immobilization of five nitro benzoic acids was performed. Addition of four different alkenyl Grignard reagents and basic cleavage leads to substituted methyl indole carboxylates in excellent purities. Features of this reaction are the stability of halide groups, ester moieties, and tolerance of o,o'-unsubstituted nitro resins. Heck and Sonogashira reactions are also possible with immobilized indoles.