Merck
  • Leukocyte Infiltration Triggers Seizure Recurrence in a Rat Model of Temporal Lobe Epilepsy.
Leukocyte Infiltration Triggers Seizure Recurrence in a Rat Model of Temporal Lobe Epilepsy.
Inflammation (2016-04-05)
Zanhua Liu, Suping Wang, Jinjie Liu, Feng Wang, Yi Liu, Yongbo Zhao
ABSTRACT
Epilepsy, which affects about 1 % of the population worldwide, leads to poor prognosis and increased morbidity. However, effective drugs providing satisfactory control on seizure relapse were rare, which encouraged more etiological studies. Whether inflammation is one of key events underlying seizure is in debate. In order to explore the role of inflammatory in the pathogenesis and development of epilepsy, we conducted intra-caudal vein injection of leukocytes to aggravated brain inflammatory process in kainic acid-induced seizure model in this study. The results showed that intravenous administration of activated leukocytes increased the frequency and reduced the latent phase of seizure recurrences in rat models of epileptic seizure, during which leukocyte inflammation, brain-blood barrier damage, and neuron injury were also significantly aggravated, indicating that leukocyte infiltration might facilitate seizure recurrence through aggravating brain inflammation, brain-blood barrier damage, and neuron injury.