Merck
  • Synthesis of N-substituted indole derivatives via PIFA-mediated intramolecular cyclization.
Synthesis of N-substituted indole derivatives via PIFA-mediated intramolecular cyclization.
Organic letters (2006-12-15)
Yunfei Du, Renhe Liu, Gregory Linn, Kang Zhao
ABSTRACT
[Structure: see text] A variety of N-arylated and N-alkylated indole derivatives were synthesized by way of a phenyliodine bis(trifluoroacetate) (PIFA)-mediated intramolecular cyclization. This novel method allows for the construction of an indole skeleton by joining the N-atom on the side chain to the benzene ring at the last synthetic step. Other novel pyrrole-fused aromatic compounds can also be achieved by this method.
Sigma-Aldrich
[Bis(trifluoroacetoxy)iodo]benzene, 97%