Merck
  • Synthesis of a novel ceramide analogue via Tebbe methylenation and evaluation of its antiproliferative activity.
Synthesis of a novel ceramide analogue via Tebbe methylenation and evaluation of its antiproliferative activity.
Organic letters (2005-04-09)
Xuequan Lu, Gilbert Arthur, Robert Bittman
ABSTRACT
[reaction: see text] A new analogue of (2S,3R)-ceramide (2) with a methylene group at C4 has been synthesized from d-tartaric acid (3) by using Tebbe methylenation as the key step. Compound 2 exhibited markedly higher antiproliferative activity on mouse embryonic fibroblast (MEF) cells than natural (2S,3R,4E)-ceramide (1).