Merck
  • PreS1 epitope recognition in newborns after vaccination with the third-generation Sci-B-Vac vaccine and their relation to the antibody response to hepatitis B surface antigen.
PreS1 epitope recognition in newborns after vaccination with the third-generation Sci-B-Vac vaccine and their relation to the antibody response to hepatitis B surface antigen.
Virology journal (2009-01-22)
Ulla B Hellström, Kazimierz Madalinski, Staffan Pe Sylvan
ABSTRACT
Sci-B-Vac is a recombinant, hepatitis B vaccine derived from a mammalian cell line and containing hepatitis B surface antigen (HBsAg) as well as preS1 and preS2 antigens. Few studies have been performed on the antibody responses to preS1 in relation to the antibody to hepatitis B surface antigen (anti-HBs) response during immunisation of healthy children with preS-containing vaccines. In this study 28 healthy newborns were randomly selected to receive either 2.5 microg or 5.0 microg of the Sci-B-Vac vaccine. Children received three doses of vaccine according to a 0-, 1-, 6-month scheme. Antibodies against the S-protein and three synthetic peptides mimicking three B-cell preS1 epitopes, (21-32 amino acid epitope), (32-47 amino acid epitope) and the C-terminal (amino acid epitope 94-117) were determined at 6 and 9 months. Fourteen (50%) of the 28 newborns had detectable levels of anti-preS1 (21-32) antibodies; 15 (54%) were anti-preS1 (32-47) reactive and 12 (43%) were anti-preS1 (94-117) reactive at 6 or 9 months after initiation of the vaccination. Significantly higher levels of anti-HBs were observed in the sera of patients with detectable anti-preS1 (32-47) reactivity (24 550 +/- 7375 IU/L, mean +/- SEM) as compared with the non-reactive sera (5991 +/- 1530 IU/L, p < 0.05). The anti-HBs levels were significantly lower if none (p < 0.05) or one (p < 0.025) of the preS1 (21-32, 32-47, 94-117) peptides were recognised compared with the anti-HBs levels if two or three peptides were recognised. Recognition of several preS1 epitopes, and in particular, the epitope contained within the second half of the hepatocyte binding site localised in the hepatitis B surface protein of the third-generation hepatitis B vaccine is accompanied by a more pronounced antibody response to the S-gene-derived protein in healthy newborns.
Product Description
Supelco
Antimony Standard for AAS, TraceCERT®, 1 g/L Sb in nitric acid (nominal concentration)