Merck
  • A high-performance liquid chromatography assay with a triazole-bonded column for evaluation of d-amino acid oxidase activity.
A high-performance liquid chromatography assay with a triazole-bonded column for evaluation of d-amino acid oxidase activity.
Biomedical chromatography : BMC (2015-07-16)
Megumi Iwasaki, Yoshiyuki Kashiwaguma, Chihiro Nagashima, Mao Izumi, Ayano Uekusa, Sumiko Iwasa, Mayu Onozato, Hideaki Ichiba, Takeshi Fukushima
ABSTRACT
Elution profiles of kynurenic acid (KYNA) and 7-chlorokynurenic acid (Cl-KYNA) were examined by high-performance liquid chromatography (HPLC) using a triazole-bonded stationary phase column (Cosmosil® HILIC) under isocratic elution of a mobile phase consisting of CH3 CN-aqueous 10 mm ammonium formate between pH 3.0 and 6.0. The capacity factors of KYNA and Cl-KYNA varied with both the CH3 CN content and the pH of the mobile phase. The elution order of KYNA and Cl-KYNA was reversed between the CH3 CN- and H2 O-rich mobile phases, suggesting that hydrophilic interactions and anion-exchange interactions caused retention of KYNA and Cl-KYNA in the CH3 CN- and H2 O-rich mobile phases, respectively. The present HPLC method using a triazole-bonded column and fluorescence detection (excitation 250 nm, emission 398 nm) was applied to monitor in vitro production of KYNA from d-kynurenine (d-KYN) by d-amino acid oxidase (DAO) using Cl-KYNA as an internal standard. A single KYNA peak was clearly observed after enzymatic reaction of d-KYN with DAO. Production of KYNA from d-KYN was suppressed by the addition of commercial DAO inhibitors. The present HPLC method can be used to evaluate DAO activity and DAO inhibitory effects in candidate drugs for the treatment of schizophrenia.
Sigma-Aldrich
3-Methylpyrazole-4-carboxylic acid, 97%