Merck
  • Design, synthesis and SAR of piperidyl-oxadiazoles as 11β-hydroxysteroid dehydrogenase 1 inhibitors.
Design, synthesis and SAR of piperidyl-oxadiazoles as 11β-hydroxysteroid dehydrogenase 1 inhibitors.
European journal of medicinal chemistry (2013-01-29)
Guangxin Xia, Xiaodi You, Lin Liu, Haiyan Liu, Jianfa Wang, Yufang Shi, Ping Li, Bing Xiong, Xuejun Liu, Jingkang Shen
ABSTRACT
The potential roles of 11β-HSD1 inhibitors in metabolic syndrome, T2D and obesity were well established and currently several classes of 11β-HSD1 inhibitors have been developed as promising agents against metabolic diseases. To find potent compounds with good pharmacokinetics, we used the bioisosterism approach, and designed the compound 2 and 3 bearing an 1,2,4-oxadiazole ring to replace the amide group in compound 1. Guided by docking study, we then transformed compound 3 into a potent lead compound 4a by changing sulfonamide group to amide. To elaborate this series of piperidyl-oxadiazole derivatives as human 11β-HSD1 inhibitors, we explored the structure-activity relationship of several parts of the lead compound. Based on their potency toward human 11β-HSD1 two compounds 4h and 4q were advanced to pharmacokinetic study. It was found that 4h and 4q are potent and selective human 11β-HSD1 inhibitors with better pharmacokinetic properties than those of the original piperidine-3-carboxamide compound 1, and suitable for further in vivo preclinical study in primate model.