Merck
  • Lack of association of glutathione S-transferase M3 gene polymorphism with the susceptibility of lung cancer.
Lack of association of glutathione S-transferase M3 gene polymorphism with the susceptibility of lung cancer.
Asian Pacific journal of cancer prevention : APJCP (2012-11-22)
Xu Feng, Chun-Qiang Dong, Jun-Jie Shi, Hua-Fu Zhou, Wei He, Bao-Shi Zheng
ABSTRACT
The conclusions of published reports on the relationship between the glutathione S-transferase M3 (GSTM3) A/B gene polymorphism and the risk of lung cancer are still debated. This meta-analysis was performed to evaluate the association between GSTM3 and the risk of lung cancer. Association investigations were identified from PubMed, Embase, and Cochrane Library, and eligible studies were included and synthesized using a meta-analysis method. Eight reports were included into this meta-analysis for the association of GSTM3 A/B gene polymorphism and lung cancer susceptibility, covering 1,854 patients with lung cancer and 1,926 controls. No association between the GSTM3 A/B gene polymorphism and lung cancer was found in this meta-analysis (B allele: OR = 1.25, 95% CI: 0.89-1.76, P = 0.20; BB genotype: OR = 1.53, 95% CI: 0.71-3.32, P = 0.28; AA genotype: OR = 0.85, 95% CI: 0.59-1.23, P = 0.39). The GSTM3 A/B gene polymorphism is not associated with lung cancer susceptibility. However, more studies on the relationship between GSTM3 A/B gene polymorphism and the risk of lung cancer should be performed in the future.
Sigma-Aldrich
GST M3-3, Recombinant Human