Merck
  • The micronucleus assay in mammalian cells in vitro to assess health benefits of various phytochemicals.
The micronucleus assay in mammalian cells in vitro to assess health benefits of various phytochemicals.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-11-02)
Roberta Meschini, Andrea Berni, Silvia Filippi, Gaetano Pepe, Maria Rosaria Grossi, Adayapalam T Natarajan, Fabrizio Palitti
ABSTRACT
We evaluated the protective effects of Gentiana lutea extracts (GLEx) and 6-Gingerol (6-G) on clastogenicity of N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) and 7,12-dimethylbenz(α) anthracene (DMBA) in vitro on HepG2 cells using the frequencies of induced micronuclei (MN) as the end point. Pre-, post- and simultaneous treatments with GLEx or 6-G and the carcinogens were carried out. Both GLEx post- and simultaneous treatments reduced the frequencies of MN induced by MNNG and DMBA. Probably this effect is due to an increase of cytostasis and a physico-chemical interaction between GLEx and DMBA under simultaneous treatment. Pre- and simultaneous treatments with 6-G significantly reduced the yield of MNNG-induced micronuclei without affecting % of cytostasis. Simultaneous treatment with 6-G plus DMBA resulted in reduction in the frequency of MN and an increase in cytotoxicity compared to sample treated alone with DMBA, whereas a post-treatment, caused a significant decrease in the yield of MN compared with DMBA alone without any cytotoxic effect. These results are compared with our earlier data obtained in the same system with other phytochemicals. It is concluded that for a critical evaluation of the protective effects of phytochemicals, both the influence on the induced MN and induced cytostasis have to be considered.
Sigma-Aldrich
[6]-Gingerol, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Pricing and availability is not currently available.
SAFC
L-Glutamine
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Pricing and availability is not currently available.