Merck
Shoc2/Sur8 protein regulates neurite outgrowth.
PloS one (2014-12-17)
Gonzalo Leon, Lucia Sanchez-Ruiloba, Andrea Perez-Rodriguez, Teresa Gragera, Natalia Martinez, Silvia Hernandez, Berta Anta, Olga Calero, Carlota A Garcia-Dominguez, Lara M Dura, Daniel Peña-Jimenez, Judit Castro, Natasha Zarich, Pilar Sanchez-Gomez, Miguel Calero, Teresa Iglesias, Jose L Oliva, Jose M Rojas
ABSTRACT
The Shoc2 protein has been implicated in the positive regulation of the Ras-ERK pathway by increasing the functional binding interaction between Ras and Raf, leading to increased ERK activity. Here we found that Shoc2 overexpression induced sustained ERK phosphorylation, notably in the case of EGF stimulation, and Shoc2 knockdown inhibited ERK activation. We demonstrate that ectopic overexpression of human Shoc2 in PC12 cells significantly promotes neurite extension in the presence of EGF, a stimulus that induces proliferation rather than differentiation in these cells. Finally, Shoc2 depletion reduces both NGF-induced neurite outgrowth and ERK activation in PC12 cells. Our data indicate that Shoc2 is essential to modulate the Ras-ERK signaling outcome in cell differentiation processes involved in neurite outgrowth.