Merck
  • Molecular basis of tubulin transport within the cilium by IFT74 and IFT81.
Molecular basis of tubulin transport within the cilium by IFT74 and IFT81.
Science (New York, N.Y.) (2013-08-31)
Sagar Bhogaraju, Lukas Cajanek, Cécile Fort, Thierry Blisnick, Kristina Weber, Michael Taschner, Naoko Mizuno, Stefan Lamla, Philippe Bastin, Erich A Nigg, Esben Lorentzen
ABSTRACT
Intraflagellar transport (IFT) of ciliary precursors such as tubulin from the cytoplasm to the ciliary tip is involved in the construction of the cilium, a hairlike organelle found on most eukaryotic cells. However, the molecular mechanisms of IFT are poorly understood. Here, we found that the two core IFT proteins IFT74 and IFT81 form a tubulin-binding module and mapped the interaction to a calponin homology domain of IFT81 and a highly basic domain in IFT74. Knockdown of IFT81 and rescue experiments with point mutants showed that tubulin binding by IFT81 was required for ciliogenesis in human cells.