Merck
  • Cytofluorometric determination of nuclear DNA in living and preserved algae.
Cytofluorometric determination of nuclear DNA in living and preserved algae.
Stain technology (1982-09-01)
H M Hull, R W Hoshaw, J C Wang
ABSTRACT
Three DNA-localizing fluorochromes used in conjunction with epi (incident) UV illumination were examined for sensitivity and selectivity for the cytofluorometric determination of nuclear DNA in ten species of six algal genera: Mougeotia, Oedogonium, Sirogonium, Spirogyra and Zygnema among the green algae, and the marine red alga Polysiphonia boldii. In comparison with absorption photometry for the determination of nuclear DNA, the cytofluorometric procedure proved to be simpler and considerably more sensitive. Following staining with 4',6-diamidino-2-phenylindole (DAPI), nuclei fluoresce blue-white, the fluorescence intensity of the DNA-DAPI complex being considerably greater than that of the unbound dye molecule. Algal strains stained with 2,5-bis[4'-aminophenyl(1')]-1,3,4-oxadiazole (BAO) also showed brilliant blue-white nuclear fluorescence. Although the BAO schedule requires the use of freshly prepared dye and sulfite water, and careful control of hydrolysis, nuclear fluorescence of the stained specimens does not fade under irradiation of the UV beam as rapidly as it does with certain other fluorochrome procedures. A more useful fluorochrome was the fungal antibiotic mithramycin. Its staining schedule is simple and the bright orange-yellow fluorescence of the nuclei is associated with an exceptional degree of sensitivity and specificity for DNA. Forty-eight-year-old preserved filaments of Spirogyra jatobae, stained with either BAO or mithramycin, exhibited a fluorescence brilliance of nuclear and chloroplast DNA equal to that of fresh specimens of this species. The three schedules, but particularly the one with mithramycin, have proven useful in providing indirect evidence for variation in ploidy level in several of the above algal genera, and in verifying the assumed ploidy level of the gametophyte (haploid) and tetrasporophyte (diploid) of Polysiphonia boldii.