Merck
  • Site-directed mutagenesis of recombinant human beta 2-glycoprotein I identifies a cluster of lysine residues that are critical for phospholipid binding and anti-cardiolipin antibody activity.
Site-directed mutagenesis of recombinant human beta 2-glycoprotein I identifies a cluster of lysine residues that are critical for phospholipid binding and anti-cardiolipin antibody activity.
Journal of immunology (Baltimore, Md. : 1950) (1996-10-15)
Y Sheng, A Sali, H Herzog, J Lahnstein, S A Krilis
ABSTRACT
beta2-Glycoprotein I (beta2GPI) is a phospholipid-binding serum protein with anticoagulant properties. It plays a vital role in the binding of anti-cardiolipin Abs purified from patients with autoimmune disease when assayed in a cardiolipin (CL) ELISA. Based on a three-dimensional model of beta2GPI, electrostatic calculations, and earlier peptide studies, a highly positively charged amino acid sequence, Lys282-Asn-Lys-Glu-Lys-Lys287, located in the fifth domain of beta2GPI, has been predicted to be the phospholipid binding site. We tested this hypothesis by site-directed mutagenesis of residues in the predicted phospholipid binding site and by assessing the mutants for phospholipid binding and anti-beta2GPI activity. A single amino acid change from Lys286 to Glu significantly decreased the binding of beta2GPI to CL. Double and triple mutants 2k (from Lys286, 287 to Glu286, 287), 2ka (from Lys284, 287 to Glu284, 287), and 3k (from Lys284, 286, 287 to Glu284, 286, 287) possessed no binding of Ab to beta2GPI in a CL ELISA, as well as no inhibitory activity on the binding of iodinated native beta2GPI to CL. These results indicate that the residues Lys284, Lys286, and Lys287 in the fifth domain of beta2GPI are critical for its binding to anionic phospholipids and its subsequent capture for binding of anti-beta2GPI Abs.