- Four-component domino strategy for the combinatorial synthesis of novel 1,4-dihydropyrano[2,3-c]pyrazol-6-amines.
Four-component domino strategy for the combinatorial synthesis of novel 1,4-dihydropyrano[2,3-c]pyrazol-6-amines.
ACS combinatorial science (2013-10-24)
Selvaraj Kanchithalaivan, Sathiyamoorthi Sivakumar, Raju Ranjith Kumar, Palani Elumalai, Qazi Naveed Ahmed, Anil K Padala
PMID24147861
ABSTRACT
An efficient one-pot four-component domino protocol for the combinatorial synthesis of novel 1,4-dihydropyrano[2,3-c]pyrazol-6-amines has been achieved. This transformation presumably occurs via cyclization-Knoevenagel condensation-Michael addition-tautomerism-intramolecular O-cyclization-elimination sequence of reactions. The significant features of this reaction include expedient one-pot process, short reaction time, no column chromatographic purification, excellent yield, and readily available starting materials.