Merck
Mutations in the deubiquitinase gene USP8 cause Cushing's disease.
Nature genetics (2014-12-09)
Martin Reincke, Silviu Sbiera, Akira Hayakawa, Marily Theodoropoulou, Andrea Osswald, Felix Beuschlein, Thomas Meitinger, Emi Mizuno-Yamasaki, Kohei Kawaguchi, Yasushi Saeki, Keiji Tanaka, Thomas Wieland, Elisabeth Graf, Wolfgang Saeger, Cristina L Ronchi, Bruno Allolio, Michael Buchfelder, Tim M Strom, Martin Fassnacht, Masayuki Komada
ABSTRACT
Cushing's disease is caused by corticotroph adenomas of the pituitary. To explore the molecular mechanisms of endocrine autonomy in these tumors, we performed exome sequencing of 10 corticotroph adenomas. We found somatic mutations in the USP8 deubiquitinase gene in 4 of 10 adenomas. The mutations clustered in the 14-3-3 protein binding motif and enhanced the proteolytic cleavage and catalytic activity of USP8. Cleavage of USP8 led to increased deubiqutination of the EGF receptor, impairing its downregulation and sustaining EGF signaling. USP8 mutants enhanced promoter activity of the gene encoding proopiomelanocortin. In summary, our data show that dominant mutations in USP8 cause Cushing's disease via activation of EGF receptor signaling.
Sigma-Aldrich
Endoproteinase Arg-C from mouse submaxillary gland, Type XX-S, lyophilized powder, 250-800 units/mg protein (Lowry)