- (E)-3-(4-Methyl-phen-yl)-1-(1,3-thia-zol-2-yl)prop-2-en-1-one.
(E)-3-(4-Methyl-phen-yl)-1-(1,3-thia-zol-2-yl)prop-2-en-1-one.
Acta crystallographica. Section E, Structure reports online (2012-06-22)
Annamalai Palaniappan, Arumugam Arulmozhivarman, Rajamanickam Ramachandran, Subramanian Srinivasan, Sivakolunthu Senthan
PMID22719636
ABSTRACT
In the title chalcone, C(13)H(11)NOS, derived from the condensation of p-tolualdehyde and 1-(1,3-thia-zol-2-yl)ethanone, the olefine group has a trans configuration. No classical hydrogen bonding is present in the crystal structure.