Merck
  • Synthesis of semiconducting polymer microparticles as solid ionophore with abundant complexing sites for long-life Pb(II) sensors.
Synthesis of semiconducting polymer microparticles as solid ionophore with abundant complexing sites for long-life Pb(II) sensors.
ACS applied materials & interfaces (2014-11-19)
Mei-Rong Huang, Yong-Bo Ding, Xin-Gui Li, YingJun Liu, Kai Xi, Chen-Long Gao, R Vasant Kumar
ABSTRACT
Intrinsically electrically semiconducting microparticles of semiladder poly(m-phenylenediamine-co-2-hydroxy-5-sulfonic aniline) structures containing abundant functional groups, like -NH-, -N=, -NH2, -OH, -SO3H as complexation sites, were efficiently synthesized by chemical oxidative copolymerization of m-phenylenediamine and 2-hydroxy-5-sulfonic aniline. The obtained copolymers were found to be nonporous spherical microparticles that were able to achieve greater π-conjugated structure, smaller particle aggregate size, and stronger interaction with Pb(II) ions than poly(m-phenylenediamine) containing only -NH-, -N=, and -NH2. A potentiometric Pb(II) sensor was fabricated on the basis of the copolymer microparticles as a crucial solid ionophore component within plasticized PVC. The sensor exhibited a Nernstian response to Pb(II) ions over a wide concentration range, together with a fast response, a wide pH range capability, a long lifetime of up to 5 months, and good selectivity over a wide variety of other ions and redox species. The process for synthesizing the microparticles and fabricating the Pb(II)-sensor can be facilely scaled-up for use in the straightforward long-term online monitoring of Pb(II) ions in heavily polluted wastewaters. This study develops an understanding of the facile synthesis of conducting microparticles bearing many functional groups and their structures governing the potentiometric susceptibility toward interaction between Pb(II) ions and the microparticles for fabricating robust long-lived Pb(II)-sensor, signifying the potential suitability of such novel materials for inexpensive sensitive detection of Pb(II) ions.
Sigma-Aldrich
m-Phenylenediamine, flakes, 99%
Pricing and availability is not currently available.