Merck
  • Efficient preparation of N-phenylsulfenyl ketimines from oximes or nitro compounds without racemization of alpha-stereocenters.
Efficient preparation of N-phenylsulfenyl ketimines from oximes or nitro compounds without racemization of alpha-stereocenters.
Organic letters (2007-10-09)
Jordi Burés, Carles Isart, Jaume Vilarrasa
ABSTRACT
As N-sulfenyl imines (e.g., RR'C=N-SAr) can be readily transformed to their N-sulfinyl imines (RR'C=N-SOAr), N-sulfonyl imines (RR'C=N-SO2Ar), and N-sulfonyl oxaziridines, the very mild procedure developed to convert ketoximes and secondary nitro derivatives to N-arenesulfenyl ketimines constitutes a new and efficient route to all these series of compounds. The configuration of the alpha-stereocenters is retained.