Merck
  • An unprecedented 3-D SHG MOF material of silver(I) induced by chiral triple helices.
An unprecedented 3-D SHG MOF material of silver(I) induced by chiral triple helices.
Chemical communications (Cambridge, England) (2013-03-02)
Huan Yang, Rui-Li Sang, Xiang Xu, Li Xu
ABSTRACT
An unprecedented 3-D SHG-active metal-organic framework (MOF) of silver(I), Ag-Amidn (1) (HAmidn: 2-amino-4,5-imidazoledicarbonitrile), has been successfully prepared by employing the new single ligand. The cooperative coordination of the imidazole and carbonitrile N atoms leads to the chiral triple helices accounting for the noncentrosymmetric space group, which are linked together to form a rare 10(3) topological structure. Ag-Amidn (1) with high thermal stability and insolubility may be considered as a potential second-order NLO material.
Sigma-Aldrich
2-Amino-4,5-imidazoledicarbonitrile, 97%