Merck
Bromidotetra-kis-(2-isopropyl-1H-imidazole-κN)copper(II) bromide.
Acta crystallographica. Section E, Structure reports online (2011-11-09)
Sylwia Godlewska, Joanna Socha, Katarzyna Baranowska, Anna Dołęga
ABSTRACT
The Cu(II) atom in the title salt, [CuBr(C(6)H(10)N(2))(4)]Br, is coordinated in a square-pyramidal geometry by four imidazole N atoms and one bromide anion that is located at the apex of the pyramid. The cations and the anions form a two-dimensional network parallel to (001) through N-H⋯Br hydrogen bonds.
Sigma-Aldrich
2-Isopropylimidazole, 98%