Merck
  • A chiral thioxanthone as an organocatalyst for enantioselective [2+2] photocycloaddition reactions induced by visible light.
A chiral thioxanthone as an organocatalyst for enantioselective [2+2] photocycloaddition reactions induced by visible light.
Angewandte Chemie (International ed. in English) (2014-03-22)
Rafael Alonso, Thorsten Bach
ABSTRACT
Thioxanthone 1, which was synthesized in a concise fashion from methyl thiosalicylate, exhibits a significant absorption in the visible light region. It allows for an efficient enantioselective catalysis of intramolecular [2+2] photocycloaddition reactions presumably by triplet energy transfer.
Sigma-Aldrich
Methyl thiosalicylate, 97%