Merck
  • Depletion of plasma albumin for proteomic analysis of Bothrops jararaca snake plasma.
Depletion of plasma albumin for proteomic analysis of Bothrops jararaca snake plasma.
Journal of biomolecular techniques : JBT (2011-07-09)
Karen de Morais-Zani, Kathleen Fernandes Grego, Aparecida Sadae Tanaka, Anita Mitico Tanaka-Azevedo
ABSTRACT
The proteomic analysis of plasma samples represents a challenge as a result of the presence of highly abundant proteins such as albumin. To enable the detection of biomarkers, which are commonly low-abundance proteins, in complex blood fluids, it is necessary to remove high-abundance proteins efficiently. Moreover, there is a range of about 10 orders of magnitude for the abundance of different protein species in serum. Here, we describe for the first time a study of reptilian albumin depletion using resins usually used in mammalian plasma depletion procedures. We performed the depletion of albumin from Bothrops jaraca plasma using the HiTrap Blue high-performance column (GE Healthcare Life Sciences, Piscataway, NJ, USA) and the kit Albumin & IgG Depletion SpinTrap column (GE Healthcare Life Sciences). In addition, proteomic approaches were used to analyze reptilian plasma. Our results showed that B. jararaca albumin bound to both columns, but those interactions were not enough to remove a large amount of albumin to reach an enrichment of low-abundance proteins. Although the depletion techniques used in this work were not the best to remove B. jararaca plasma albumin, our present work highlights the similarity between B. jararaca and mammalian albumin, contributing to the knowledge of comparative hemostatic proteins.