- A case of eosinophilic pneumonia and vasculitis induced by diflunisal.
A case of eosinophilic pneumonia and vasculitis induced by diflunisal.
Chest (1997-06-01)
M W Rich, R A Thomas
PMID9187209
ABSTRACT
Drug-induced pneumonitis is an uncommon complication of nonsteroidal anti-inflammatory drug administration. Herein is the first reported case of pneumonitis resulting from diflunisal therapy. The patient demonstrated clinical and biopsy evidence of systemic vasculitis. She responded dramatically to administration of systemic glucocorticoids.
MATERIALS