Merck
  • n-Doping of organic electronic materials using air-stable organometallics.
n-Doping of organic electronic materials using air-stable organometallics.
Advanced materials (Deerfield Beach, Fla.) (2011-11-08)
Song Guo, Sang Bok Kim, Swagat K Mohapatra, Yabing Qi, Tissa Sajoto, Antoine Kahn, Seth R Marder, Stephen Barlow
ABSTRACT
Air-stable dimers of sandwich compounds including rhodocene and (pentamethylcyclopentadienyl)(arene)ruthenium and iron derivatives can be used for n-doping electron-transport materials with electron affinities as small as 2.8 eV. A p-i-n homojunction diode based on copper phthalocyanine and using rhodocene dimer as n-dopant shows a rectification ratio of greater than 10(6) at 4 V.