Merck
(2-Methyl-1-phenyl-sulfonyl-1H-indol-3-yl)methanol.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
G Chakkaravarthi, V Dhayalan, A K Mohanakrishnan, V Manivannan
ABSTRACT
In the title compound, C(16)H(15)NO(3)S, the plane of the phenyl ring forms a dihedral angle of 80.37ā€…(8)Ā° with the indole ring system. The crystal packing is stabilized by weak O-Hā‹ÆO hydrogen bonds which link the mol-ecules into infinite chains along the a axis of the crystal.
Sigma-Aldrich
2-Methylindole-3-carboxaldehyde, 97%