Merck
  • Phosphine-catalyzed [4 + 3] annulation for the synthesis of highly functionalized bicyclo[3.2.2]nonadienes.
Phosphine-catalyzed [4 + 3] annulation for the synthesis of highly functionalized bicyclo[3.2.2]nonadienes.
Organic letters (2009-08-13)
Suqing Zheng, Xiyan Lu
ABSTRACT
A phosphine-catalyzed [4 + 3] annulation of modified allylic carbonates with methyl coumalate was developed. This strategy offered a powerful method for the construction of bicyclo[3.2.2]nonadiene skeleton with high stereoselectivity.
Sigma-Aldrich
Methyl coumalate, 98%