Merck
  • Derivatization and template-guided ligation of oligodeoxyribonucleotides using cyanogen bromide and N-substituted morpholines.
Derivatization and template-guided ligation of oligodeoxyribonucleotides using cyanogen bromide and N-substituted morpholines.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2001-09-07)
Z A Shabarova, O A Fedorova, N G Dolinnaya, M B Gottikh
ABSTRACT
Cyanogen bromide has been found to induce the template-guided condensation of oligonucleotides only in the presence of N-substituted morpholines. Based on 31P, 1H and 13C NMR spectroscopy data, the mechanism of the phosphomonoester group activation by cyanogen bromide in N-substituted morpholine buffers is suggested. It has also been shown that BrCN can be used for the synthesis of oligonucleotide derivatives in aqueous solution.