Merck
  • Baking properties and biochemical composition of wheat flour with bran and shorts.
Baking properties and biochemical composition of wheat flour with bran and shorts.
Journal of the science of food and agriculture (2013-07-31)
Leonid Kaprelyants, Sergey Fedosov, Dmytro Zhygunov
ABSTRACT
Bran, being a by-product of grain grinding, is characterised by a high biological value and is thus widely used in food production. In this study, different streams of bran and shorts from the wheat graded milling process were incorporated into wheat flour at levels of 5, 11, 17 and 23% (w/w) to investigate their influence on the nutritional and baking properties of flour. Bran and shorts streams improved the baking properties of flour blends. The best result in the case of graded flour blends with different bran products was obtained at the 95:5 ratio. The products containing peripheral parts of grain had higher proteolytic enzyme and superoxide dismutase activities and lower trypsin inhibitor content and β-amylase activity compared with graded flour. Streams of wheat milled fractions including peripheral parts of grain increase the content of bioactive substances and dietary fibre in blends with wheat graded flour.
Sigma-Aldrich
β-Amylase from barley, Type II-B, 20-80 units/mg protein (biuret)
Sigma-Aldrich
β-Amylase from sweet potato, Type I-B, ammonium sulfate suspension, ≥750 units/mg protein (E1%/280)
Sigma-Aldrich
β-Amylase from sweet potato