- Inhibitors of acyl-CoA:cholesterol O-acyltransferase (ACAT) as hypocholesterolemic agents: synthesis and structure-activity relationships of novel series of sulfonamides, acylphosphonamides and acylphosphoramidates.
Inhibitors of acyl-CoA:cholesterol O-acyltransferase (ACAT) as hypocholesterolemic agents: synthesis and structure-activity relationships of novel series of sulfonamides, acylphosphonamides and acylphosphoramidates.
Bioorganic & medicinal chemistry letters (1999-01-01)
H T Lee, W H Roark, J A Picard, D R Sliskovic, B D Roth, R L Stanfield, K L Hamelehle, R F Bousley, B R Krause
PMID9871672
ABSTRACT
Sulfoacetic acid, phosphoramidate, and phosphoramide analogs of the ACAT inhibitors, CI-999 and CI-1011 were synthesized. The structure-activity relationships of these compounds as ACAT inhibitors are described.