Merck
  • The isolation of secondary metabolites and in vitro potent anti-cancer activity of clerodermic acid from Enicosanthum membranifolium.
The isolation of secondary metabolites and in vitro potent anti-cancer activity of clerodermic acid from Enicosanthum membranifolium.
Bioorganic & medicinal chemistry (2007-04-03)
Mai Efdi, Tomohiro Itoh, Yukihiro Akao, Yoshinori Nozawa, Mamoru Koketsu, Hideharu Ishihara
ABSTRACT
Four compounds were isolated from Enicosanthum membranifolium. The structures of the compounds were confirmed by spectroscopic data. Their structures were determined as N-trans-feruloyltyramine, R-(-)-mellein, clerodermic acid, and salicifoline chloride as a quaternary alkaloid compound. The structures of R-(-)-mellein and salicifoline chloride were confirmed by using X-ray diffraction. Clerodermic acid was shown to induce potent apoptosis against human leukemia HL60 cells.
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)