- Selective synthesis of 1-O-alkyl(poly)glycerol ethers by catalytic reductive alkylation of carboxylic acids with a recyclable catalytic system.
Selective synthesis of 1-O-alkyl(poly)glycerol ethers by catalytic reductive alkylation of carboxylic acids with a recyclable catalytic system.
ChemSusChem (2012-09-25)
Marc Sutter, Wissam Dayoub, Estelle Métay, Yann Raoul, Marc Lemaire
PMID23001817
ABSTRACT
(Poly)glycerol monoethers were synthesized in good yield and selectivity by the catalytic reductive alkylation of glycerol, diglycerol, and triglycerol with readily available, cheap and/or bio-sourced carboxylic acids. The reaction was catalyzed by 1 mol % of Pd/C under 50 bar H(2) using an acid ion-exchange resin as a recyclable cocatalyst. The catalytic system was recycled several times, and a mechanism is proposed for this transformation.