Merck
  • On the role of the bridging dicyanamidobenzene ligand in a new binuclear ruthenium complex: [{Ru(tpy)(thd)}2(mu-dicyd)][PF6] with tpy = 2,2':6',2' '-terpyridine and thd = 2,2,6,6-tetramethyl-3,5-heptanedione.
On the role of the bridging dicyanamidobenzene ligand in a new binuclear ruthenium complex: [{Ru(tpy)(thd)}2(mu-dicyd)][PF6] with tpy = 2,2':6',2' '-terpyridine and thd = 2,2,6,6-tetramethyl-3,5-heptanedione.
Inorganic chemistry (2006-11-07)
Muriel Fabre, Joël Jaud, Mohamed Hliwa, Jean-Pierre Launay, Jacques Bonvoisin
ABSTRACT
The dicyanamidobenzene-bridge diruthenium complex [{Ru(tpy)(thd)}(2)(mu-dicyd)][PF(6)] ([3][PF(6)]) (dicyd = 1,4-dicyanamidobenzene, tpy = 2,2':6',2' '-terpyridine, thd = 2,2,6,6-tetramethyl-3,5-heptanedione) and its mononuclear counterpart [Ru(tpy)(thd)(Ipcyd)] (2) [Ipcyd = 4-iodophenylcyanamide anion (Ipcyd(-))] were synthesized and fully characterized. Cyclic voltammetry of 3 showed the presence of four reversible one-electron redox couples. UV-vis-NIR spectroelectrochemistry and EPR spectroscopy of the electrogenerated paramagnetic intermediates were used to ascertain the oxidation-state distribution. The stable starting dinuclear complex 3(+) is found to be a ligand-centered anion radical as shown by EPR spectroscopy, magnetic susceptibility measurements, and DFT calculations. Oxidation of 3(+) to 3(2+) led to an EPR silent system due to substantial intramolecular antiferromagnetic interaction of the electron spins carried by the low spin ruthenium(III) atom and the bridging anion radical dicyanamido (dicyd(*)(-)), an observation which was supported by UV-vis-NIR, X-ray structure, and DFT calculations. Complex 3(3+) presented an EPR spectra consistent with a total effective spin S = (1)/(2) issued from an antiferromagnetic interaction of electron spins carried by two low spin ruthenium(III) atoms and the bridging anion radical dicyd(*)(-) in accordance with UV-vis-NIR. This study shows that the dicyanamidobenzene bridging ligand has indubitably a noninnocent behavior.
Sigma-Aldrich
2,2,6,6-Tetramethyl-3,5-heptanedione, 98%