Merck
  • Synthesis of (2S,2'R,3S,4R)-2-(2'-hydroxy-21'-methyidocosanoylamino)-1,3,4-pentadecanetriol, the ceramide sex pheromone of the female hair crab, Erimacrus isenbeckii.
Synthesis of (2S,2'R,3S,4R)-2-(2'-hydroxy-21'-methyidocosanoylamino)-1,3,4-pentadecanetriol, the ceramide sex pheromone of the female hair crab, Erimacrus isenbeckii.
Bioscience, biotechnology, and biochemistry (2002-09-13)
Yui Masuda, Masao Yoshida, Kenji Mori
ABSTRACT
The ceramide sex pheromone [(2S,2'R,3S,4R)-2-(2'-hydroxy-21-methyldocosanoylamino)-1,3,4-pentadecanetriol (1)] of the female hair crab (Erimacrus isenbeckii) was synthesized by starting from (S)-serine and 12-bromo-1-dodecanol.